Check the following items before you calibrate the instrument using the Auto-Ready hardware.
- The API source is properly installed.
- The Auto-Ready enclosure door is closed.
Perform the following, as applicable:
- Run the New Vial Installed diagnostic if a new Auto-Ready (FlexMix) vial is installed. This diagnostic checks for any common problems from leaks, clogs, and so on.
- From the Diagnostics pane, navigate to Hardware → Auto-Ready → Fluidics.
- Run the Check Spray Stability and Check Calibrant Solution diagnostics after the spray has been established.
- From the Diagnostics pane, navigate to Hardware → Auto-Ready.
NOTE
You can run the Start Spray diagnostics to manually start the Auto-Ready spray (independent of running calibrations) if there are issues with the spray. Once the diagnostic starts, the Auto-Ready hardware automatically attempts to establish a stable flow to the sprayer and applies a high voltage to the sprayer to generate calibrant ions.
From the Diagnostics pane, navigate to Hardware → Auto-Ready.
TIP
You can stop the Auto-Ready spray at any time by running the Stop Spray diagnostic.
The following figure shows the Diagnostics pane with the Auto-Ready diagnostics options.