Check the helium gas and nitrogen gas regulators to determine how much gas is in each tank.
CAUTION
Before you operate the MS, verify that there is sufficient nitrogen for the API source. Do not operate the MS without nitrogen gas flow as this can damage the source and front end optics due to particulate matter in the atmosphere.
If the system runs out of nitrogen gas while you are acquiring scans in the Tune application, a prompt appears with a warning that the nitrogen supply is probably empty. If you are running an acquisition sequence, the data system completes the current run. The system does not proceed to the next sample because the MS cannot return to the Ready state when the nitrogen set point cannot be reached. Meanwhile, the MS continues scanning.
The following table describes the procedures for checking the gas supplies.
Procedures for Checking the Gas Supplies
Gas | Procedure |
---|---|
UHP Helium gas | Procedure
|
Nitrogen gas | Procedure
If necessary, adjust the pressure using the tank pressure regulator. |
When operating 24 hours and 7 days a week, the system typically consumes the following amounts of gas per day:
|