The MS contains an Auto-Ready™ ion source that is used exclusively to calibrate the instrument. The Auto-Ready components are located behind the small access door on the instrument front panel and include the following:
  • Auto-Ready pump
  • Calibration solution vial and vial receptacle
  • Flow sensor and flow sprayer
NOTE
The instrument ships with an empty calibration solution vial in the Auto-Ready vial receptacle. Replace the empty vial with a new prefilled (unopened) FlexMix vial before you calibrate the instrument using Auto-Ready. See Replace the Auto-Ready Calibration Solution Vial.
The following figure shows the Auto-Ready enclosure.
Figure Auto-Ready enclosure
123456
No.DescriptionNo.Description
1
Nitrogen gas feed line
4
Plunger assembly plate
2
Flow sensor attached to holding bracket
5
Vial locking lever in the locked position
3
PEEK fittings
6
Seated calibration solution vial
The components come preassembled at the factory and are inspected by the Thermo Fisher Scientific service engineer during the installation. The following items are inspected:
  • The gas fitting on the right-side of the pump manifold, which inputs pressurized nitrogen to the pump during operation.
  • The gas fittings on the center of pump manifold and the plunger assembly, which transmit regulated nitrogen from the manifold to the calibration solution vial.
  • The gas fitting on the left-side of the pump manifold, which provides a bleed path for gas out of the manifold.
  • The fitting on the top of the plunger, which seals the PEEK tubing where the calibration solution exits the vial.
  • The fitting on the bottom-side of the flow sensor, which transmits liquid from the vial to the sensor.
  • The fitting on the top-side of the flow sensor, which transmits liquid from the sensor to the flow sprayer.
  • The flow sprayer, which needs to be locked in the proper position on the right side of the vacuum manifold.