A holding bracket provides a secure location to mount the syringe pump and the optional divert/inject valve on the MS. Follow this procedure to assemble the optional holding bracket and to mount the syringe pump and the divert/inject valve.
Optional Tool
- #2 Phillips screwdriver
Procedure
- Attach the bottom shelf to the holding bracket using the provided screws.
- Place the divert/inject valve on the bottom shelf. Then, attach the top shelf using two screws.
- Place the syringe pump on the angled support bracket.
- The bottom of the syringe pump must be centered with the spring-type screw on the angled support bracket.
- Tighten the spring-type screw to secure the syringe pump to the angled support bracket using your fingers or a screwdriver.
- Place the holding bracket assembly (with the syringe pump and divert/inject valve) on the top-left surface of the MS.