You can configure (plumb) the divert/inject valve as a loop injector or as a divert valve. The loop injector configuration is used for flow injection analysis.
The following table provides details on the loop injector configuration.
Loop Injector Configuration | Description |
---|---|
In the first position (also known as the load position), the following occurs:
| |
In the second position (also known as the inject position), the following occurs:
NOTE If you load sample into the inlet port when the valve is in the inject position, the sample flows to waste. |
When using the divert valve configuration, you can switch the solvent front, gradient endpoint, or any portion of the LC run to flow to waste.
The following table provides details on the divert valve configuration.
Divert Valve Configuration | Description |
---|---|
In the first position (also known as the detector position), the flow from the LC pump enters the valve through port 2, exits port 1, and flows to the API source. | |
In the second position (also known as the waste position), the following occurs:
|
NOTE
For more information on the various sample introduction techniques and plumbing, see To Connect the Inlet Plumbing.