The optional external Rheodyne™ MX Series II™ divert/inject valve is a 6-port motorized valve that can switch between two positions. The ports on the valve use the standard fitting (10-32 type) for high-pressure and 1/16 inch outer diameter (OD) tubing.
The following table shows the internal flow paths for the divert/inject valve positions.
NOTE
In the Method Editor, the valve positions are identified as 1–2 (port 1 to port 2) and 1–6 (port 1 to port 6).
Valve position | Description |
---|---|
In the first position (1–2), the ports are internally connected as follows:
In the second position (1–6), the valve rotates clockwise one position and connects the ports internally as follows:
|
The divert/inject valve can be plumbed as a loop injector for flow injection analysis or as a divert valve. For more information about the divert/inject valve, see the following topics: