The external Chemyx™ Fusion 100T2 syringe pump delivers sample solution from the syringe to the API source. A motorized pusher block depresses the syringe plunger at the flow rate specified in the data system. You can also start, stop, and prime the syringe pump using the data system. The syringe pump can be installed a holding bracket (not provided), which provides a secure location to mount the syringe pump and the optional divert/inject on the MS.
You can use the syringe and syringe pump in the following ways:
  • To directly infuse sample solution into the API source
  • To infuse sample into the solvent stream that is produced by an LC pump
  • To automatically load sample into an optional divert/inject valve
NOTE
The typical syringe pump flow rate for calibration is 3–5 µL/min.
For more details about the syringe pump, refer to the manufacturer's manual.
Figure Chemyx Fusion 100T2 syringe pump
Chemyx Fusion 100T2 syringe pump