To open the Tune Help or Method Editor Help, do the following as applicable:
Procedure
- Tune Help: From the Tune application, select Options
and then select Tune Help.
- Method Editor Help: From the Xcalibur data system (Roadmap view), select Instrument Setup and then navigate to the top menu bar and select Help → Stellar Help.
NOTE
To open context-sensitive help from either Help system, press the F1 key from within a specific view, page, or dialog box.