In the Tune preferences, you can specify the system alert to display warnings, recovered errors and warnings, and information message types. If these items are not selected, the Alert Console does not display these message types.
The following figure shows the Alert Console Options in the Tune Preferences window.
Figure Alert Console Options in the Tune preferences
Alert Console Options in the Tune preferences
Procedure
  1. Open the Tune application.
  2. Select Options and then select Preferences.
  3. In the Alerts Console Options area, specify the alert settings as applicable.
NOTE
All error alerts are displayed in the Alerts Console window by default and cannot be disabled.