Use the Ion Source pane to specify the ion source parameters. Real-time colored indicators identify the readback status for each parameter and are as follows:
Green: Indicates that the parameter is within its expected range.
Orange: Indicates that the parameter is trending toward the expected range.
Red: Indicates that the parameter is outside of its expected range.
The following figure shows the colored indicators next to various parameters.