The Diagnostics pane is typically used by a Thermo Fisher Scientific service engineer. It provides item-specific instrument calibration parameters. As a standard user, you can access limited parameters to run specific, per-item, calibrations.
NOTE
Contact Thermo Fisher Scientific Technical Support if you need assistance with diagnostics.
Prerequisites
  1. Select the Options icon and then select Preferences to open the Tune Preferences window.
  2. Under Report Options, specify your preferred report settings.
Procedure
  1. Select the Diagnostics icon on the lower-right corner of the Tune window to open the Diagnostics pane.
  2. In the Diagnostics pane, select Check Only if you want to run the check procedures only on the Standard Calibration category.
  3. Select the arrow next to the category that you want to expand.
  4. Select the components that you want to test.
  5. Select Start.
For more information on generating reports, see Generate Reports.