The Calibration pane contains an instructions area that provides a quick set of manual calibration instructions. These instructions are only viewable when the ESI/H-ESI ion source type is detected on the MS.
Figure Instructions area in the Calibration pane
Instructions area in the Calibration pane
NOTE
Thermo Fisher Scientific recommends calibrating the instrument using the internal Auto-Ready calibration hardware.