The Calibration pane contains a Calibration area where you can check and calibrate the MS. The Tune application automatically enforces dependencies between certain instrument calibrations, ensuring that each aspect of instrument performance is checked, calibrated, or both only after its dependencies have been checked, calibrated, or both. If any particular item fails its check, then any items that depend on that failed calibration are skipped.
The following table provides details on the parameters that are used to check and calibrate your instrument.
Item | Description |
---|---|
Mode | Specifies the calibration mode. The options for this parameter are as follows:
|
Polarity | Specifies the ion polarity mode (positive or negative). |
NOTE
- For more details on how to calibrate your MS, see Calibration.
- To generate a calibration report, see Generate Reports.