The dimensions and approximate weight of the instrument are as follows:
Width | Depth | Height | Weight |
---|---|---|---|
66 cm | 81 cm | 70 cm | 142 kg |
26 in. | 32 in. | 28 in. | 314 lb |
NOTE
The MS can be placed on a laboratory workbench or table. If you use a table, ensure that it can support twice the weight of the MS and the weight of any additional installed hardware components, such as an ion source, syringe pump, divert/inject valve, or optional FAIMS interface.