To avoid excess liquid buildup in the API source housing when you increase the liquid flow rate from the LC system, adjust the following parameters: gas flow rates (sheath, auxiliary, and sweep), ion transfer tube temperature, and vaporizer temperature.
NOTE
The recommended start settings for the spray voltage (Positive 3,500 volts and Negative 2,500 volts ) are independent of the LC flow rate.
The following table lists the recommended start settings for operating a system using a H-ESI configured API source with a range of LC solvent flow rates. The range for the Current LC Flow in the application is 0–3,000 µL/min.
LC Flow Rate (μL/min) | ≤14.99 | 15–99.99 | 100–199.99 | 200–499.99 | 500–999.99 | 1,000–3,000 |
---|---|---|---|---|---|---|
Sheath Gas (Arb) | 5 | 25 | 35 | 50 | 60 | 75 |
Aux Gas (Arb) | 2 | 5 | 7 | 10 | 15 | 25 |
Sweep Gas (Arb) | 0 | 0 | 0 | 1 | 2 | 2 |
Ion Transfer Tube | 325 | 325 | 325 | 325 | 380 | 400 |
Vaporizer Temp (°C) | 20 | 75 | 275 | 350 | 350 | 400 |