You can check the recommended API source settings for various LC flow rates using the Tune application or Method Editor.
Procedure
- Do one of the following:
- Tune application: Select Stellar Tune
on the desktop to open the Tune application. Then, select the Ion Source tab.
- Method editor: Open Xcalibur and select Instrument Setup. Then, select the Global Parameters tab.
- In the Ion Source pane of the Tune application or the Ion Source Properties pane of the Method Editor, enter the LC flow rate in the Current LC Flow (μL/min) box. Then, select Get Defaults.