The instrument application programming interface (iAPI) allows you to write custom applications in C# that can modify instrument parameters in real-time. You can generate a license using the Ardia Platform Link application or by sending a request to info.iapi@thermofisher.com. Documentation, example code, and compiled libraries are available at https://github.com/thermofisherlsms/iapi.
For information on the Ardia Platform Link and iAPI license, refer to the Ardia Platform Link Setup Guide at docs.thermofisher.com.