Special notices, symbols, and caution labels appear in boxes. Those pertaining to safety also have corresponding symbols. Some symbols are also marked on the instrument itself and can appear in color or in black and white.
Notice, symbol, or label | Description |
---|---|
IMPORTANT | Highlights information necessary to prevent damage to software, loss of data, or invalid test results; or might contain information that is critical for optimal performance of the product. |
NOTE | Highlights information of general interest. |
TIP | Highlights helpful information that can make a task easier. |
Caution: Read the cautionary information associated with this task. | |
| Chemical hazard: Observe safe laboratory practices and procedures when handling chemicals. Only work with volatile chemicals under a fume or exhaust hood. Wear gloves and other protective equipment, as appropriate, when handling toxic, carcinogenic, mutagenic, corrosive, or irritant chemicals. Use approved containers and proper procedures to dispose of waste oil and when handling wetted parts of the instrument. |
| Heavy object: The physical handling of this instrument requires a team effort to lift or move. This instrument is too heavy and bulky for one person alone to handle safely. |
| Hot surface: Before touching the atmospheric pressure ionization (API) source assembly, give the heated components time to cool. |
| Pinch point: Keep hands away from the specified areas. |
| Risk of electric shock: This instrument uses voltages that can cause electric shock and/or personal injury. Before servicing, shut down the instrument and disconnect it from line power. While operating the instrument, keep covers on. |
| Risk of eye injury: Eye injury can occur from splattered chemicals, airborne particles, or sharp objects. Wear safety glasses when handling chemicals or servicing the instrument. |
| Sharp object: Avoid handling the tip of the syringe needle. |
| Trip obstacle: Be aware of cords, hoses, or other objects located on the floor. |