The Tune application includes a system alert feature that provides real-time notifications for various events such as errors and warnings. The Alerts Console, which is minimized by default is displayed at the bottom of the Tune window when system alerts are detected after the launch of the Tune application. The Alerts Console displays icons indicating the count for each alert type. If system alerts are not detected on the system, the console is not displayed.
The following figure shows an example of the Alerts Console minimized at the bottom of the Tune window.
Figure Alerts Console minimized
12345
No.DescriptionNo.Description
1
Error alert
4
Information alert
2
Warning alert
5
Minimized Alert Console at the bottom of the Tune window
3
Recovered errors and warnings alert
You can expand the minimized Alerts Console by selecting an area near the count icons. This opens the alert details within a docked window, as shown in the following figure.
Figure Alerts Console in the docked view
123
No.DescriptionNo.Description
1
Event table
3
Maximize button
2
Alert Console (docked view)
To switch from the docked view to a floating window, select the Maximize button located on the right-side of the docked view. In a floating window, the Alerts Console displays the full event table and the control options, as shown in the following figure.
Figure Alerts Console in a floating window
12
No.DescriptionNo.Description
1
Alert Console control options
2
Event table
The event table in the Alert Console can display a maximum limit of 1,000 alerts. Error and Warning alerts remain in the view until they are recovered. Information alerts will drop off from the view once the 1,000 line limit is reached. You can organize and sort events by toggling the column type at the top of the table header.