The Stellar MS is part of the Thermo Scientific family of mass spectrometers. The key components are:
- OptaMax™ Plus ion source
- Two mass analyzers
- Auto-Ready™ ESI source that automates the calibration process
- Dry forepump
- Syringe pump for the infusion of user samples, if needed
- (Optional) modular divert/inject valve