Use the Hybrid View layout to create experiments consisting of scan types and optional filters types. This is workflow node-centric layout that offers a wide range of adjustable parameters that appear in the right-side object properties pane of all scans and filters.
The first node of a workflow tree is the master scan. The master scan is the parent scan in a data-dependent experiment, or it can be the only scan in the experiment.
Figure Hybrid View layout
12345
No.DescriptionNo.Description
1
Hybrid view icon
4
Workflow node-centric layout area
2
Scan types
5
Drag and drop a scan type to the "Place Scan Here" node
3
Optional filter types
NOTE
Elements in the workflow node-centric layout are referenced as icons, objects, and nodes.