The Quan View is a table-centric layout where the scan parameters appear in a table format. This layout has fewer experiment parameters compared to the Hybrid view. The scan parameters can be in adjusted from within the table or from the additional parameters that appear in the right-side properties pane.
Figure Quan View layout
1234
No.DescriptionNo.Description
1
Quan View icon
3
Table-centric layout area
2
Scan types
4
Drag and drop a scan type to the "Place Scan Here" node