Select the Scan Parameters tab to open Scan Parameters page. On this page, you can create an LC/MS instrument method that consists of one or more experiments. Each experiment has its own timeline and workflow. You can also build a workflow by selecting application-specific experiment templates.
NOTE
The Method Timeline and Settings area are displayed at the top of the Global Parameters and Scan Parameters page.
The following figure describes the main areas of the Scan Parameters page (also known as the workflow editor).
Figure Scan Parameters page
7123456
No.
Description
No.
Description
1
Scan types
5
Experiment workflow area
2
View toolbar (Quan View, Hybrid View, and Templates View)
6
Settings
3
Scan Parameters tab
7
Preferences icon
4
Method timeline