When you open the Method Editor, the Global Parameters page appears by default. On this page, you can configure the settings for the ion source, syringe pump, divert valves, and any connected devices controlled by a contact closure signal. The Method Editor uses these settings for all scans.
Use the Method Timeline area to create a timeline for your experiment. Each timeline is a graphical representation of the experiment’s specified time.
NOTE
The Method Timeline and Settings area are displayed at the top of the Global Parameters and Scan Parameters page.
The following figure describes the main areas of the Global Parameters page.
Figure Global Parameters page
2341
No.
Description
No.
Description
1
Select to configure the Ion Source, Syringe, Divert valve, and Output Contact Closure parameters
3
Method timeline
2
Global Parameters tab
4
Settings