The Method Editor can be used to create and set up experiments by using the entire complement of scan types, advanced filters, and conditional logic. You create an MS instrument method (combined with the optional autosampler and liquid chromatograph instrument methods) by defining the experiment type and setting various parameters. These include settings for the MS, syringe pump, and divert valve; and the mass ranges and fragmentation transitions for the experiments.
NOTE
For detailed information on method parameter settings, refer to the Method Editor Help.
For more information, see these topics: