The Status pane displays real-time instrument readback information for various components in the MS. The colored status readback indicators update every few seconds and are as follows:
  • Green: Indicates that the parameter is within its expected range.
  • Orange: Indicates that the parameter is trending toward the expected range.
  • Red: Indicates that the parameter is outside of its expected range.
The following figure displays an example of the Status pane.
Figure Status pane
123
No.DescriptionNo.Description
1
Colored status readback indicators
3
Status pane button
2
By Board or By Function selection
Procedure
  1. If the Status tab, History tab, and Favorites tab are not visible on the right of the window, select the expand icon to show the tabs.
  2. Select Status to open the Status pane.
  3. From the dropdown list in the title bar of the Status pane, do one of the following:
  4. - Select By Board to display a list of components based on the electronic PCB board that controls their operation.
  5. - Select By Function to display a list of components based on their functionality in the MS.