The MP3 ion optics transmit ions from the ICRM to the LIT. The MP3 ion optics include the MP3 multipole.
The MP3 multipole is an array of circular rods that act as an ion transmission device. The MS applies an RF voltage to the rods, generating an electric field that guides the ions along the axis of the multipole.