The MS uses six DC axial trapping voltages, one for each rod section on both cells. These voltages establish axial trapping by creating potential wells. These DC axial trapping voltages allow the mass analyzer to perform its storage and scan-out functions.