The MS contains a dual-cell, two-dimensional linear ion trap (LIT) mass analyzer. The LIT consists of a front trap lens (TL1), a high pressure cell, a center trap lens (TL2), a low pressure cell (analytical trap), and a back trap lens (TL3). The front, center, and back trap lenses are metal plates with a circular hole in the center through which the ion beam passes. These trap lenses provide conductance limits.
Figure Dual-pressure linear ion trap assembly
Dual-pressure linear ion trap assembly
Figure Dual-pressure linear ion trap assembly (expanded view)
1234567
No.DescriptionNo.Description
1
Top PCB
5
Front trap lens (TL1)
2
Low-pressure cell (analyzer trap)
6
Holder assembly
3
Center trap lens (TL2)
7
Back trap lens (TL3)
4
High-pressure cell
The mass analyzer cell is comprised of an array of four hyperbolic rods arranged around a central axis. The rods are segmented into three sections and have narrow slits for ion ejection. Ions are ejected only through slots in the X-rods during scan-out.
Figure Hyperbolic rods in the mass analyzer cell
123YZX
No.DescriptionNo.Description
1
Front section
3
Back section
2
Center section
For more information. see these topics: