The solenoid-operated vent valve vents the vacuum manifold with filtered air. The vent valve is closed when the solenoid is energized. The vent opens when the MS no longer receives external power, as with a power failure or when you turn off the main power switch.