The spray cone is an integral component of the API source interface. The ion transfer tube fits through a small orifice in the center of the spray cone. A graphitized Vespel™ O-ring acts as a seal between the spray cone and the back of the ion transfer tube’s nose cone.
Figure Ion transfer tube, O-ring, and spray cone
123
No.DescriptionNo.Description
1
High-capacity ion transfer tube
3
Spray cone
2
Vespel O-ring