The vacuum manifold, a thick-walled, aluminum chamber, encloses the API source interface, ion optics, mass analyzers, and ion detection system. It has multiple removable top cover plates, various electrical feedthroughs, and gas inlets.
For details on the typical pressure readbacks, see Check the Vacuum Pressure Levels.