The vacuum system evacuates the region around the API source interface, ion optics, mass analyzers, and ion detection system. The inlet gas hardware controls the flow of the collision gas, sheath gas, auxiliary gas, and air (during venting) into the MS.
for more information, see these topics: