Flush the syringe and the inlet components (that is, the sample transfer line, sample tube, and spray insert) after completing the calibration procedures and also before you change from one analyte solution to another.
For more information, see these topics: