The following figure shows the Calibration pane with Auto-Ready.
Figure Calibration pane with Auto-Ready
123456
No.DescriptionNo.Description
1
Colored status indicator
4
Recommended calibration date and last successful calibration date
2
Calibration tab
5
FlexMix volume status
3
Polarity status toggle
6
Calibration options (Mode and Polarity)
Prerequisites
  1. Check the FlexMix volume from the Calibration pane. A minimum volume of 5% (approximately 250 µL) is required to perform an instrument calibration. The system prevents starting a calibration routine when the minimum volume is below that value.
Procedure
  1. Open the Tune application. For instructions, see Tune Application Window.
  2. Select the Options icon and then select Preferences.
  3. In the Current Calibration Options area, select Auto-Ready as the calibration source.
  4. (Optional) In the System Self-Check Options area, select the checkbox to enable the system self-check option to schedule periodic instrument calibration checks.
  5. Select OK to exit the Tune Preferences dialog box.
  6. The system automatically sets FlexMix as the calibration solution.
  7. From the Tune application, select the Calibration pane.
  8. In the Calibration area, select one of the following options, as applicable:
  9. Mode:
  10. - Check, Calibrate if required (recommended)
  11. - Calibrate
  12. Polarity: Positive or Positive and Negative
  13. Select Start.
  14. The calibration operation begins with the selected options. After the system has completed a successful calibration, the Calibration pane resets to show the date of the next recommended calibration for each calibration set.
NOTE
Thermo Fisher Scientific recommends that you include a shutdown method at the end of every sequence that uses an LC workflow when scheduling Auto-Ready calibrations with the Self-Check feature. The Self-Check process automatically adjusts system settings, such as the source gases, temperature settings, and so on.
Include the following actions in the shutdown method:
  • Set the vaporizer temperature to 0 °C
  • Set the LC flow rate to under 10 µL/min
  • Set source gases to minimal levels, which helps reduce source housing dryness