Run the calibration from the Tune application, starting with positive polarity and then continuing with negative polarity using the FlexMix calibration solution (P/N A39239).
Prerequisites
- Load a clean 500 µL syringe with at least 250uL of the FlexMix calibration solution.
- Set up the Plumbing for Direct Infusion
- Set up the MS for Calibration
- Establish a Stable Ionization Spray
Procedure
- Open the Tune application.
- Select the Options
icon and then select Preferences.
- The Tune Preferences dialog box appears.
- In the Current Calibration Options area, select Other as the calibration source. Then select OK to close the dialog box.
- From the Tune application, select the Calibration pane.
- In the Calibration area, select one of the following options as applicable:
- Mode:
- - Check, Calibrate if required (recommended)
- - Calibrate
- Polarity: Positive or Negative
- Select Start.
- The calibration operation begins. After the system has completed a successful calibration, the status area in the Calibration pane resets to show the date of the next recommended calibration for each calibration set.
- Repeat this procedure in negative ion mode.
IMPORTANT
Before you start using your analyte, follow the procedure in To Flush the Inlet Components After Calibration.