To automatically inject a set of samples, connect an LC system with an autosampler to the API source. Use the autosampler to inject sample solution into the flow from an LC pump. In a typical LC/MS experiment, direct the solvent flow through an LC column to separate the compounds of a mixture before they are directed into the API source.
Follow the appropriate procedure based on your system configuration: