The instrument can use two ESI modes, which have different system functions as follows:
- Heated-electrospray ionization (H-ESI) mode used for chemical analysis. For more information, see H-ESI Mode for Analysis.
- Electrospray ionization (ESI) mode used by the low-flow ESI spray insert and the Auto-Ready calibration hardware. For more information, see Calibration.