The power module, which is located on the right side of the MS and the power mode options (On/Standby/Off) that are available from the Tune application and data system control the on/off status of various MS components.
The following figure shows the power module which contains the Electronics switch and Main Power switch.
- Electronics switch in the Service Mode position: This turns off all components in the MS, except the +24 V power supply, forepump, turbomolecular pump, and cooling fans.
- Main Power switch in the Off position: This removes all power to the MS, including the vacuum system.
The following table summarizes the on/off status of MS components, voltages, and API gas flows under varying power conditions.
On/Off Status of MS Components, Voltages, and API Gas Flows
MS Component | Power Mode Options | Power Module | |||
---|---|---|---|---|---|
On | Standby | Off1) | Electronics (Service Mode) | Main Power (Off) | |
Vent valve | Closed | Closed | Closed | Closed | Open |
APCI corona discharge needle | On | Off | Off | Off | Off |
Conversion dynode | On | Off | Off | Off | Off |
Electron multiplier | On | Off | Off | Off | Off |
ESI needle spray voltage | On | Off | Off | Off | Off |
Helium gas (collision gas)2) | On | On | Off | Off | Off |
Power supply | On | Off | Off | Off | Off |
Vaporizer temperature | On | Off | Off | Off | Off |
Ion transfer tube DC offset | On | On | Off | Off | Off |
Ion transfer tube temperature control | On | On | Off | Off | Off |
Mass analyzer, DC offset voltage | On | On | Off | Off | Off |
Mass analyzer, RF voltage | On | On | Off | Off | Off |
MP00 and MP0 ion optics, DC offset voltage | On | On | Off | Off | Off |
MP00 and MP0 ion optics, RF voltage | On | On | Off | Off | Off |
RF lens voltage | On | On | Off | Off | Off |
Embedded computer | On | On | On | Off | Off |
Sheath, auxiliary, and sweep gases 3) | On | On | On | Off | Off |
Convection vacuum gauge (collision cell) | On | On | On | Off | Off |
Ionization vacuum gauge (mass analyzer) | On | On | Off | Off | Off |
Cooling fans | On | On | On | On | Off |
Forepump | On | On | On | On | Off |
Convectron™ vacuum gauge (foreline) | On | On | On | Off | Off |
Turbomolecular pump and controller | On | On | On | On | Off |
Vent delay PCB | On | On | On | On | Off |