You can reset the MS if it is not communicating with the data system computer by using the reset button. The button is located in the communications panel on the right side of the MS, as shown in the following figure.
Figure Reset button on the communication panel
Prerequisites
  1. The MS and data system computer are on and operational.
Procedure
  1. Hold down the reset button for 3 seconds.
The following sequence of events occurs:
  • The embedded computer restarts. All LEDs on the front panel turn off except the Power LED.
  • After several seconds, the Communication LED turns yellow and then green to indicate that the MS and the data system are communicating. The data system transfers operational software to the MS.
  • After a few minutes, the System LED turns yellow to indicate that the software transfer from the data system to the MS is complete and that the MS is in Standby mode. When you change the mode from Standby to On, the System LED turns green to indicate that the MS is functional.