Check the solvent level in the solvent waste container daily. If necessary, empty the container and dispose of the solvent waste according to your local regulations.
Check the solvent level in the solvent waste container daily. If necessary, empty the container and dispose of the solvent waste according to your local regulations.