You can manually control the divert/inject valve using the control buttons on the front of valve enclosure. First, take the valve out of Remote mode by pressing the Remote button. The Remote LED will become unlit and the position arrows will become active.
  • When the valve is in the divert valve configuration, you can divert the LC flow between the MS and waste.
  • When the valve is in the loop injector configuration, you can switch between the load and inject positions.
Figure Divert/inject valve (front view)
123
No.DescriptionNo.Description
1
Valve position indicator
3
Valve control buttons
2
6-port, two-position valve