The ion optics focus the ions that are produced in the API source and transmits them to the ion detection system. The following figure shows a schematic of the Stellar MS ion optics.
Figure Stellar MS ion optics
31245678910
No.
Description
No.
Description
1
Sweep cone
6
Quadrupole mass analyzer
2
Ion transfer tube
7
Ion Concentrating Routing Multipole (ICRM) collision cell
3
RF lens
8
MP3 ion optics
4
MP00 ion optics
9
Dual-pressure linear ion trap (mass analyzer)
5
MP0 ion optics with neutrals blocker
10
Photomultiplier