The exterior cover panels on the instrument contain the necessary components for the instrument connections and operation. For a description of the panels, see the following topics:
Front Panel
The front of the instrument contains the OptaMax Plus ion source, a built-in drain system, and LEDs that indicate the status of the instrument.
1234
No.DescriptionNo.Description
1
Built-in drain system
3
LED indicators
2
OptaMax Plus ion source
4
Auto-Ready enclosure access door
Left-side Panel
The left-side panel contains the vacuum, gas, and solvent waste connections.
Right-side Panel
The right-side panel contains a recessed area with the communications panel and the power entry module.
12
No.DescriptionNo.Description
1
Communications panel (hidden from this view)
2
Power entry module
Rear Panel
The rear of the instrument contains grates that cover the cooling fans. The filter for the cooling fans is located behind the front panel.