The MS receives line power through the power entry module located on the right-side panel (with respect to the front face view) of the MS.
CAUTION
In an emergency, do not use the Electronics switch to turn off power to the MS. Instead, place the Main Power switch in the Off position and disconnect the power cord from the electrical outlet.
Power Entry Module Item Descriptions
Item | Description |
---|---|
Main Power |
|
Electronics switch |
|
V65 Pump Enable | The Pump Enable connector provides a connection to the forepump. The MS can start and stop the forepump by using a relay control cable that connects to the forepump’s contact closure connector. |
Power In | Power plug receptacle (230 Vac, 50/60 Hz, 10 A Max). |
AC Output | AC output receptacle (230 Vac, 50/60 Hz, 2.0 A Max). This receptacle is reserved for future use. |