The following table describes the LED indicators on the front panel.
LED Indicator Descriptions
LED | State | Description |
---|---|---|
Power | Green | (The Electronics switch is in the Operating Mode position.) The MS is receiving power. |
Off | (The Electronics switch is in the Service Mode position.) The MS is not receiving full power for operation. CAUTION Line power (220 Vac) to the MS is still present and there are high voltages within certain areas of the instrument. | |
System | Green | The MS is in On mode and scanning. |
Yellow | The MS is in Standby mode. | |
Off | The MS is in Off mode. | |
Vacuum | Green | The vacuum is within the allowable operating range. |
Yellow | The vacuum is outside the allowable operating range. | |
Off | The MS is either powered off or in the process of starting up. | |
Communication | Green | The MS and data system are communicating. |
Yellow | The MS and data system are trying to establish a communication link. | |
Off | (The Electronics switch is in the Service Mode position) The MS is in Off mode. | |
Scan | Flashing blue | The MS is in On mode and scanning. |
Off | The MS is either in Standby mode or Off mode and not scanning. |