Use the Thermo Foundation Instrument Configuration to configure the instrument, if applicable.
Prerequisites
- Install the Stellar 1.0 QF1 software.
Procedure
- From the Windows taskbar, open the Thermo Foundation Instrument Configuration by navigating to the following path:
- Start > Thermo Scientific Foundation 3.1> Instrument Configuration
- Highlight the Stellar icon, then select Add to place it in the Configured Devices list.
Figure Thermo Foundation Instrument Configuration window - Highlight the Stellar icon and select Configure, or double-click on the icon to open the Instrument Configuration dialog box.
- Specify the configuration parameters for the MS, as applicable.
- For details on the instrument configuration parameters, refer to the Tune Help.
- Select OK to close the Instrument Configuration window.
- Select Done to close the Thermo Foundation Instrument Configuration window.
- Restart the instrument computer if prompted.