The minimum hardware and software system requirements are as follows:
System
Requirements
Computer
  • 3.1 GHz Intel™ Quad-Core processor
  • 16 GB RAM (32 GB recommended)
  • 1 TB hard drive
  • Two Ethernet gigabit network ports
  • DVD-ROM drive
  • Monitor display resolution of 1920 × 1080
Software
  • Microsoft™ Windows™ 10 Enterprise LTSC 2019
  • Thermo Scientific™ software:
    • Foundation 3.1 SP9
    • Xcalibur™ 4.7
    • FreeStyle™ 1.8 SP2 QF1