The Chromeleon setup program (Install.exe) accepts command-line options that can be used to trigger or fine-tune certain installation and configuration tasks and properties.
The following table shows the available command-line options.
Option
Description
/l <path/file>, /log <path/file>
Logs the installation events to the specified file/location (default location is the %TEMP% folder.)
/q, /quiet, /s, /silent
The installer runs an installation without displaying a user interface. No prompts, messages, or dialog boxes display to the user.
/passive
The installer displays a progress bar to the user that indicates that an installation is in progress but displays no prompts or error messages to the user.
/repair
Repairs the product (or installs if not installed).
/uninstall
Uninstalls the product.
/norestart
The installer never restarts the computer after the installation.
/forcerestart
The installer restarts the computer after every installation.
ResponseFile=<path\file>
Specifies the response file to be used. It's recommended that this option be combined with /passive or /quiet.
InstallFolder=<path\file>
Specifies the installation folder.
The default installation folder is C:\Program Files\Thermo\ChromeleonLink\.