Procedure
  1. Open the Chromeleon Administration Console.
  2. Select Domain Resources > Computers.
  3. Select the computer on which the WebAPI Service should be installed.
  4. Select Set Maintenance Window.
  5. Select a time frame for the installation to occur.
  6. Enable the Enforce Maintenance Window option.
  7. From the Download Software Update menu, select the package.
  8. Enable the Install Update option.
  9. Select OK and save the changes.
  10. Wait until the installation is completed.
TIP
You can find the installation log file at C:\ProgramData\Dionex\Chromeleon\Log\ChromeleonLink-Setup.log on the target computer.